Invitrogen™ eBioscience™ Human C1q Platinum ELISA Kit
For the quantitative detection of human C1q.
Manufacturer:  Invitrogen™ LSBMS2099FIVE
 View more versions of this product
Catalog No. 501125331
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human C1q
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
ELISA
0.08ng/mL
Serum, Plasma: 10μL (1:100 prediluted), CCS 50μL
C1q
5 x 96 tests
Serum, Plasma, Cell Culture Supernatant
Human
complement component 1, q subcomponent, A chain el, complement component 1, q subcomponent, alpha poly
Documents