missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Bub3 (aa 180-326) Control Fragment Recombinant Protein

Catalog No. RP102165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP102165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP102165 Supplier Invitrogen™ Supplier No. RP102165
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82108 (PA5-82108. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Specifications

Accession Number O43684
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9184
Name Human Bub3 (aa 180-326) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Aa2-050; AU019800; AU021329; AU043350; AW146323; bub3; BUB3 budding uninhibited by benzimidazoles 3 homolog; BUB3 mitotic checkpoint protein; BUB3, mitotic checkpoint protein; BUB3L; Bub3-PA; Bub3-PB; budding uninhibited by benomyl; budding uninhibited by benzimidazoles 3 homolog; budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae); C78067; CG7581; CG7581-PA; CG7581-PB; dBUB3; Dmbub3; Dmel\CG7581; Dmel_CG7581; hBUB3; mitotic checkpoint component; Mitotic checkpoint protein BUB3; OTTHUMP00000020682; testicular tissue protein Li 27; WD repeat type I transmembrane protein A72.5
Common Name Bub3
Gene Symbol BUB3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YQTRCIRAFPNKQGYVLSSIEGRVAVEYLDPSPEVQKKKYAFKCHRLKENNIEQIYPVNAISFHNIHNTFATGGSDGFVNIWDPFNKKRLCQFHRYPTSIASLAFSNDGTTLAIASSYMYEMDDTEHPEDGIFIRQVTDAETKPKSP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less