Please login to your online account to display your discounted pricing

eBioscience™ Human BMP-2, Recombinant Protein

Human Recombinant Protein

$187.05 - $419.05

Specifications

Name BMP-2
Recombinant Yes
Conjugate Unlabeled
Purity >97%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018524

 
ebioscience™
14850762
5μg Each for $187.05

5018525

 
ebioscience™
14850780
25μg Each for $419.05
Description & Specifications

Specifications

Name BMP-2
Recombinant Yes
Conjugate Unlabeled
Purity >97%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Cytokine Bioassays
Molecular Weight 13kDa
Storage Requirements Store at less than or equal to -70°C.

BMP-2 is one of fifteen bone morphogenetic proteins (BMP) belonging to the TGF-β superfamily. The BMPs were first identified as the active factors in demineralized bone matrix, with transcripts later being discovered in many other types of tissue. They are synthesized as large precursor molecules that must be cleaved into their active form and dimerize in order to exhibit functional activity. These dimers may include two identical or distinct BMP chains. BMP responsiveness appears to be limited to multipotent and immature cells. The BMPs are essential for osteogenesis and organogenesis during embryonic development, and also play a role in fracture and wound healing in adults.