Please login to your online account to display your discounted pricing

eBioscience™ Human anti-Annexin V Platinum ELISA Kit

For the quantitative detection of antibodies to human Annexin V (Vac- alpha).

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym anti Annexin A5, anti Annexin 5
Target Annexin V
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with recombinant Annexin V
HRP-Conjugate recombinant Annexin V
Human anti-Annexin V Standard lyophilized, 800ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018160

 
life technologies
LSBMS247
96 tests Each for $520.84

5018161

 
life technologies
LSBMS247TEN
10 x 96 tests Each for $2,932.34
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym anti Annexin A5, anti Annexin 5
Target Annexin V
Conjugate HRP
Sample Type Cell culture supernatant, serum
Sensitivity 1.2ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.