Please login to your online account to display your discounted pricing

Human Angiopoietin-2 Quantikine ELISA Kit, R&D Systems

$662.25 - $3,178.57

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 285
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Saliva
Sample Volume Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Saliva (50μL)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DANG20

 
r & d systems
RDSDANG20
1 Plate Each for $662.25

SANG20

 
r & d systems
RDSSANG20
6 Plates Each for $3,178.57
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 285
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Saliva
Sample Volume Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Saliva (50μL)
Sensitivity 21.3pg/mL
Species Human
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate

Angiopoietins, Ang-1, Ang-2, and Ang-3 (mouse)/Ang-4 (human), are natural agonists or antagonists of the Tie-2 receptor tyrosine kinase and are important modulators of angiogenesis. Two domains characterize the angiopoietin family of proteins: an N-terminal coiled-coil domain that mediates homo-oligomerization, and a C-terminal fibrinogen-like domain that binds Tie-2.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Saliva (50μL)
Sensitivity: 21.3pg/mL
Specificity: Natural and recombinant human Angiopoietin-2
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.