Please login to your online account to display your discounted pricing

eBioscience™ Human Adiponectin Platinum ELISA Kit

For the quantitative detection of human Adiponectin.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL (1:5000 prediluted)
Synonym ACDC, apM1, GBP-28, ADIPOQ
Target Adiponectin
Sample Type Cell culture supernatant, serum, plasma (citrate, heparin)
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Adiponectin
Biotin-Conjugate anti-human Adiponectin monoclonal antibody
Streptavidin-HRP
Human Adiponectin Standard lyophilized, 30ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125260

 
life technologies
LSBMS20322
96 tests Each for $651.05

501125261

 
life technologies
LSBMS20322TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL (1:5000 prediluted)
Synonym ACDC, apM1, GBP-28, ADIPOQ
Target Adiponectin
Sample Type Cell culture supernatant, serum, plasma (citrate, heparin)
Sensitivity 0.012ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.