missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Human Adiponectin Platinum ELISA Kit

For the quantitative detection of human Adiponectin.

Manufacturer:  Invitrogen™ LSBMS20322TEN

 View more versions of this product

Catalog No. 501125261



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Adiponectin
Biotin-Conjugate anti-human Adiponectin monoclonal antibody
Streptavidin-HRP
Human Adiponectin Standard lyophilized, 30ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

ELISA
0.012ng/mL
50μL (1:5000 prediluted)
Adiponectin
10 x 96 tests
Cell culture supernatant, serum, plasma (citrate, heparin)
Human
ACDC, apM1, GBP-28, ADIPOQ
Documents