missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Human Activin A, Recombinant Protein, Carrier-Free

Human Recombinant Protein

$825.05 - $2,057.55

Specifications

Name Activin A
Recombinant Yes
Conjugate Unlabeled
Purity ≥95%
Purification Method SDS-PAGE
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015638 Life Technologies
LS34899382
100μg Each for $825.05
5015639 Life Technologies
LS34899385
500μg Each for $2,057.55
Specifications

Description

Activin A is a member of the TGF-β superfamily, along with the other activin proteins and the structurally related inhibins. The activins are dimeric proteins formed by the association of two of the four existing β subunits, βA, βB, βC, and βE. These subunits are shared with the inhibin proteins, which are heterodimers of one β and one α chain. Activin A is a homodimer of two 13 kDa βA subunits, which can also associate with βB or βC to form Activin AB or AC, respectively. Activin A is expressed by various cell types and exhibits pleiotropic effects including the regulation of metabolism, homeostasis, cell proliferation and differentiation, apoptosis, and tissue healing. Activin A has been recently reported to work in synergy with TGF-β to promote the induction of antigen specific iTreg cells leading to inhibition of Th1 and Th2-mediated responses.
Description & Specifications

Specifications

Activin A
Unlabeled
SDS-PAGE
Cytokine Bioassays
Store at less than or equal to -70°C.
Yes
≥95%
Insect Cells
26kDa