3M™ Versaflo™ S-Series Hood and Head Covers Assemblies
Hood with reusable suspension providing head, face, neck and shoulder coverage. The 3M Versaflow S-Series Hood Assembly is one-size-fits-all.
$146.00 - $450.59
Specifications
Color White
Size Standard
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
19-805-619
View Documents
3M™
S60510
Replacement Hood with Inner Collar
Case of 10 Each for $450.59
19-805-601
View Documents
3M™
7000002347
Premium suspension hood assembly
Each for $146.00
Description
Description
Manufactured from general purpose cost effective material, with knitted inner collar. The reusable suspension hoods can be a highly cost effective for some high disposal rate environments: just the soiled hood fabric is replaced, while the suspension and air ducting can be reused. It also has an externally-adjustable airflow control that allows wearers to modify the airflow distribution to suit personal comfort needs.
  • Hood with reusable suspension providing head, face, neck and shoulder coverage
  • Provides eye and face protection to EN166—liquid splash and low energy impact (grade F)—the same classification for impact resistance as safety spectacles
  • Externally adjustable airflow to suit the wearers needs
  • Highly cost effective for high disposal rate environments
  • General purpose, cost effective fabric, with knitted collar
  • Easy Disposal - components can be separated
  • Replaceable over-hoods
Specifications
Specifications
White
Standard
Polypropylene
SDS