Please login to your online account to display your discounted pricing

HTMT dimaleate, Tocris

H1 / H2 agonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material HTMT dimaleate
Assay Percent Range >99%
Molecular Formula C19H25F3N4O·2C4H4O4
Formula Weight 614.57
Synonym 6-[2-(4-Imidazolyl)ethylamino]-N-(4-trifluoromethylphenyl)heptanecarboxamide dimaleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

064610

 
r & d systems
RDS064610
10mg Each for $177.37

064650

 
r & d systems
RDS064650
50mg Each for $746.48
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 614.57
Assay Percent Range >99%
CAS 195867-54-0
Recommended Storage Desiccate at Room Temperature
Synonym 6-[2-(4-Imidazolyl)ethylamino]-N-(4-trifluoromethylphenyl)heptanecarboxamide dimaleate
Chemical Name or Material HTMT dimaleate
Molecular Formula C19H25F3N4O·2C4H4O4
Safety and Handling

Desiccate at Room Temperature