HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications. Ideal for high-throughput PCR product cleanup using liquid handling platforms and multi-channel pipettes.
$62.35 - $7,171.70
Specifications
Storage Requirements Store at −20°C. Stable for one month at 4°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124992 Affymetrix™
LS785951PK
5,760 reactions, 12 × 8-tube strips in a tray Each for $7,171.70
501124991 Affymetrix™
LS785951EA
480 reactions, 8-tube strip Each for $632.20
501124994 Affymetrix™
LS785951000RXN
1,000 reactions, 2mL Each for $1,247.00
501124995 Affymetrix™
LS7859520RXN
20 reactions Each for $62.35
501124996 Affymetrix™
LS785954PK
23,040 reactions, 48 × 8-tube strips in a tray N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
501124997 Affymetrix™
LS785955000RXN
5,000 reactions, 10mL Each for $5,404.15
Specifications
Description
  • Mixture of Exonuclease I and Shrimp Alkaline Phosphatase for the enzymatic removal of excess primers and dNTPs following a PCR reaction
  • One-tube PCR cleanup
  • Shortened 14-minute protocol
  • Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
Description & Specifications
Specifications
Store at −20°C. Stable for one month at 4°C