Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications. Ideal for high-throughput PCR product cleanup using liquid handling platforms and multi-channel pipettes.
Manufacturer:  Applied Biosystems™ LS785951EA
 View more versions of this product
Catalog No. 501124991
Description
Description
  • Mixture of Exonuclease I and Shrimp Alkaline Phosphatase for the enzymatic removal of excess primers and dNTPs following a PCR reaction
  • One-tube PCR cleanup
  • Shortened 14-minute protocol
  • Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
Specifications
Specifications
480 reactions, 8-tube strip
Store at −20°C. Stable for one month at 4°C
Documents