missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HSPD1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00003329B01P
Description
Sequence: MLRLPTVFRQMRPVSRVLAPHLTRAYAKDVKFGADARALMLQGVDLLADAVAVTMGPKGRTVIIEQSWGSPKVTKDGVTVAKSIDLKDKYKNIGAKLVQDVANNTNEEAGDGTTTATVLARSIAKEGFEKISKGANPVEIRRGVMLAVDAVIAELKKQSKPVTTPEEIAQVATISANGDKEIGNIISDAMKKVGRKGVITVKDGKTLNDELEIIEGMKFDRGYISPYFINTSKGQKCEFQDAYVLLSEKKISSIQSIVPALEIANAHRKPLVIIAEDVDGEALSTLVLNRLKVGLQVVAVKAPGFGDNRKNQLKDMAIATGGAVFGEEGLTLNLEDVQPHDLGKVGEVIVTKDDAMLLKGKGDKAQIEKRIQEIIEQLDVTTSEYEKEKLNERLAKLSDGVAVLKVGGTSDVEVNEKKDRVTDALNATRAAVEEGIVLGGGCALLRCIPALDSLTPANEDQKIGIEIIKRTLKIPAMTIAKNAGVEGSLIVEKIMQSSSEVGYDAMAGDFVNMVEKGIIDPTKVVRTALLDAAGVASLLTTAEVVVTEIPKEEKDPGMGAMGGMGGGMGGGMFSpecifications
HSPD1 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_002156.4 | |
HSPD1 | |
HSPD1 (NP_002147.2, 1 a.a. ∼ 573 a.a) full-length human protein. | |
RUO | |
3329 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunofluorescence,Western Blot | |
Unconjugated | |
heat shock 60kDa protein 1 (chaperonin) | |
CPN60/GROEL/HLD4/HSP60/HSP65/HuCHA60/SPG13 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |