missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HSD17B10, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00003028B01P
Description
Sequence: MAAACRSVKGLVAVITGGASGLGLATAERLVGQGASAVLLDLPNSGGEAQAKKLGNNCVFAPADVTSEKDVQTALALAKGKFGRVDVAVNCAGIAVASKTYNLKKGQTHTLEDFQRVLDVNLMGTFNVIRLVAGEMGQNEPDQGGQRGVIINTASVAAFEGQVGQAAYSASKGGIVGMTLPIARDLAPIGIRVMTIAPGLFGTPLLTSLPEKVCNFLASQVPFPSRLGDPAEYAHLVQAIIENPFLNGEVIRLDGAIRMQPSpecifications
HSD17B10 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_004493.2 | |
HSD17B10 | |
HSD17B10 (NP_004484.1, 1 a.a. ∼ 261 a.a) full-length human protein. | |
RUO | |
3028 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunofluorescence,Western Blot | |
Unconjugated | |
hydroxysteroid (17-beta) dehydrogenase 10 | |
17b-HSD10/ABAD/CAMR/DUPXp11.22/ERAB/HADH2/HCD2/MHBD/MRPP2/MRX17/MRX31/MRXS10/SCHAD/SDR5C1 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |