missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HPX, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00003263B01P
Description
Sequence: MARVLGAPVALGLWSLCWSLAIATPLPPTSAHGNVAEGETKPDPDVTERCSDGWSFDATTLDDNGTMLFFKGEFVWKSHKWDRELISERWKNFPSPVDAAFRQGHNSVFLIKGDKVWVYPPEKKEKGYPKLLQDEFPGIPSPLDAAVECHRGECQAEGVLFFQGDREWFWDLATGTMKERSWPAVGNCSSALRWLGRYYCFQGNQFLRFDPVRGEVPPRYPRDVRDYFMPCPGRGHGHRNGTGHGNSICAVAHISpecifications
HPX | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
BC005395.1 | |
HPX | |
HPX (AAH05395.1, 1 a.a. ∼ 254 a.a) full-length human protein. | |
RUO | |
3263 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
hemopexin | |
- | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |