Thermo Scientific™ HPLC Syringes for CTC Instruments
$130.48 - $168.80
Specifications
Connection Type Fixed
For Use With (Equipment) CTC instruments
Gauge 22 ga
Length (English) Needle 1.9 in.
Length (Metric) Needle 50mm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Type Volume (Metric) Price Quantity  
36-5DL-991 Thermo Scientific™
365DL991
HPLC Syringe 10μL Each for $130.48
36-5HL-331 Thermo Scientific™
365HL331
Gastight™ HPLC Syringe 100μL Each for $168.80
Specifications Description & Specifications
Specifications
Fixed
22 ga
50mm
CTC instruments
1.9 in.
Each