missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ HPLC-purified dNTPs

Meet high quality standards to give excellent performance in a variety of molecular biology applications.

$86.50 - $86.50

Specifications

Storage Requirements Store at -20°C. Avoid freeze/thaw cycles
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Concentration Price Quantity  
Catalog Number Mfr. No. Quantity Concentration Price Quantity  
AB-0196 Thermo Scientific™
AB0196
1mL 20mM (5mM of each dNTP) Each for $86.50
Specifications

Description

Thermo Scientific™ HPLC-purified deoxyribonucleotide triphosphates (dNTPs) meet high quality standards to give excellent performance in a variety of molecular biology applications.

  • Available in a range of concentrations and packaging configurations for increased assay flexibility
  • AB-0196: Single vial (5mM) of combined dNTPs (dATP, dCTP, dGTP, and dTTP)
  • AB-1124: Single vial (25mM) of combined dNTPs
  • AB-0241: Four separate vials (10mM each) for each dNTP
  • AB-0315: Four separate vials (100mM each) for each dNTP

Recommended for:

PCR, RT-PCR, DNA labeling, Site-directed mutagenesis.

Description & Specifications

Specifications

Store at -20°C. Avoid freeze/thaw cycles