Please login to your online account to display your discounted pricing

HPI 1, Tocris™

Inhibits Hedgehog signaling

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material HPI 1
Assay Percent Range >95%
Molecular Formula C27H29NO6
Formula Weight 463.52
Synonym 1,4,5,6,7,8-Hexahydro-4-(3-hydroxyphenyl)-7-(2-methoxyphenyl)-2-methyl-5-oxo-3-quinolinecarboxylic acid 2-methoxyethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

383910

 
r & d systems
RDS383910
10mg Each for $221.01

383950

 
r & d systems
RDS383950
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 463.52
Quantity 10mg
Chemical Name or Material HPI 1
Assay Percent Range >95%
Molecular Formula C27H29NO6
CAS 599150-20-6
Synonym 1,4,5,6,7,8-Hexahydro-4-(3-hydroxyphenyl)-7-(2-methoxyphenyl)-2-methyl-5-oxo-3-quinolinecarboxylic acid 2-methoxyethyl ester
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C