Honeywell Safety Products™ Single-Use Max™ Ear Plugs
NRR: 33dB
Manufacturer:  Honeywell Safety Products™ MAX5H5
 View more versions of this product
Catalog No. 19-063-072
Description
Description
  • Contoured design easier to insert, resists tendency to back out of ear canal
  • Polyurethane foam enhances comfort, especially for long-term wear
  • Smooth, soil-resistant closed cell-foam skin prevents dirt build-up
  • Bell shape delivers maximum in-ear comfort
Compliance
Attenuation tested in accordance with ANSI™ S3.19-1974.
Specifications
Specifications
ANSI S3.19-1974
Uncorded
For work situations that demand a high degree of comfort, frequent changes, or where hygiene presents a problem for re-use
Polyurethane Foam
Resealable polybag
Bell
Single-use earplugs
Coral
Yes
In-ear
33 dB
5 Pairs/bag
23.63 mm
Regular