Please login to your online account to display your discounted pricing

Howard Leight™ Max™ Small Single-Use Ear Plugs

Polyurethane foam earplug offers superior noise-blocking performance in a disposable earplug

$26.53 - $26.53

Specifications

Certifications/Compliance ANSI S3.19-1974
Color Coral
Product Type Howard Leight™ Max™ Small Earplug
Size Universal
For Use With (Application) Smooth, soil resistant closed cell-foam skin prevents dirt build-up
View More Specs
Products
Catalog Number Mfr. No. Style Price Quantity    

19-021-231

 
north safety products
MAX1S
Uncorded Pack of 2000 Each for $26.53
Case of 10 Pack for $241.20
Description & Specifications

Specifications

Certifications/Compliance ANSI S3.19-1974
Color Coral
Product Type Howard Leight™ Max™ Small Earplug
Size Universal
For Use With (Application) Smooth, soil resistant closed cell-foam skin prevents dirt build-up
Material Polyurethane Foam
Noise Reduction Range (NRR) 30dB
Shape Bell

Enhanced contoured shape is easier to insert and resists backing out of the ear canal, while its softer foam delivers superior long-wearing comfort, delivering solid performance that your workers can depend on.

  • 20% smaller version of flagship Howard Leight MAX earplug, ideal for workers with smaller ear canals
  • Bell shape delivers maximum in-ear comfort
  • Contoured design easier to insert, resists tendency to back-out of ear canal
  • Smooth, soil-resistant closed cell-foam skin prevents dirt build-up
  • Bulk refills for Leight Source 400 and Source 500 Ear Plug Dispensers save time, waste and space
  • NRR: 30