Honeywell™ Howard Leight™ Leightning™ T3 Headband Ear Muff
Manufacturer:  Honeywell™ 1013941
Catalog No. 29-635-5760
Description
Description
  • Exclusive padded foam headband and super soft ear cushions eliminate pressure on head
  • Air Flow Control™ technology delivers optimal attenuation across all frequencies
  • Robust steel-wire construction withstands demanding use
  • Snap-in ear cushions replace easily
  • Telescoping height adjustment offers personalized comfort
Specifications
Specifications
Fluorescent green
Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel
30dB
Ear Muff
No
Reflective padded PUR-E, PVC headband
Over-the-head
Documents