Please login to your online account to display your discounted pricing
Howard Leight™ Leightning L2 Ear Muffs
$381.31 - $381.31
Specifications
Certifications/Compliance ANSI S3.19-1974
Product Type Ear Muff
For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel
Noise Reduction Rating (NRR) 27dB
View More Specs
Products
Catalog Number Mfr. No. Color Style Price Quantity    
29-635-5761
north safety products
1013942
Green Folding Case of 20 Each for $381.31
Description & Specifications
Specifications
Certifications/Compliance ANSI S3.19-1974
Product Type Ear Muff
For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel
Noise Reduction Rating (NRR) 27dB