Honeywell™ Howard Leight™ Leightning L2 Ear Muffs
Manufacturer: Honeywell™ 1013942
Specifications
ANSI S3.19-1974 | |
Yes | |
Ear Muffs | |
27 dB |
Green | |
Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel | |
Folding |