Honeywell™ Howard Leight™ Fusion™ Ear Plugs
Fusion™ advanced manufacturing technology delivers a combination of comfort and easy handling. Detachable nylon cord system can be used in a variety of applications.
Manufacturer:  Honeywell™ FUS30HP
 View more versions of this product
Catalog No. 19-100-154
Description
Description
  • Firm core makes insertion and removal easy
  • Constructed of soft outer flanges thermally fused to a firm inner core
  • Patented quadruple SoftFlange™ design offers excellent attenuation
  • FlexiFirm™ core makes it simple to achieve a proper fit that stays in place
  • Fabric cord detaches easily to alternate between corded and uncorded applications
Specifications
Specifications
Yes
Blue, yellow
No
Facility Maintenance, Grinding, Electrical, Welding, Demolition, Painting, Blasting, Assembly, Cleaning
Pocket-size case
HearPack case
Blue, yellow
Corded
Flanged
Class AL
Nylon
Reusable
No
No
100 Pairs/Pk.
TPE
27 dB
Regular
SDS
Documents
Accessories