missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Howard Leight™ Clarity™ C3 Ear Muffs

Improve worker safety by blocking noise, while voice and signal frequencies can be heard more naturally

$31.73 - $31.73

Specifications

Color Metallic Blue
Product Type Ear Muffs
For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, metal fabricationPetrochemicalSteelMiningTransportationUtility/Waste Management
Material Plastic
Noise Reduction Rating (NRR) 27 dB
View More Specs

Products 1
Catalog Number Mfr. No. Style Price Quantity  
Catalog Number Mfr. No. Style Price Quantity  
29-635-3559
View Documents
Honeywell™
1011146
Over-the-head
Each for $31.73
Case of 10 Each for $299.60
 
Description

Description

  • Sound management technology enhances on-the-job communication through uniform attenuation
  • Noise Reduction Rating: 27dB
  • Blocks noise while voice, signal, alarm and other high frequencies can be heard more naturally, with less distortion, combating overprotection and worker isolation
  • Robust non-deforming dielectric construction withstands use and abuse, while protecting in electrical environments
  • Height adjustment remains fixed during wear
  • Snap-in ear cushions make replacement quick and easy
  • Non-deforming outer headband provides better comfort, minimizes pressure on head, withstanding rough treatment in the toughest workplaces
  • Inner-ventilated headband provides better positioning, minimizes pressure on the head

Specifications

Specifications

Metallic Blue
Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, metal fabricationPetrochemicalSteelMiningTransportationUtility/Waste Management
27 dB
Ear Muffs
Plastic
SDS