Howard Leight™ Clarity™ C1 Earmuffs
Provide uniform attenuation that allows speech frequencies to be heard with less distortion
$91.10 - $91.10
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
29-635-3557 North Safety Products
1011142
Case of 10 Each for $91.10
Specifications
Description
  • Sound management technology
  • Enhances on-the-job communication through uniform attenuation
  • Noise Reduction Rating: 20dB
  • Clarity blocks noise while voice, signal, alarm and other high frequencies can be heard more naturally, with less distortion, combating overprotection and worker isolation
  • Dielectric/plastic construction withstands use and abuse, while protecting in electrical environments
  • Quick-click height adjustment remains fixed during wear
  • Snap-in ear cushions make replacement quick and easy