Please login to your online account to display your discounted pricing
Hourglass Industries™ HandPRO™ Series 1550 Clean Class 12 in. Vinyl Gloves
Suitable for ISO 5 and up
$179.47 - $179.47
Specifications
Certifications ASTM, FDA
Length (English) 12 in.
Length (Metric) 30.5cm
Material Vinyl
Glove Cuff Style Beaded/Rolled
View More Specs
Products
Catalog Number Mfr. No. Width (English) Width (Metric) Size Price Quantity    
19423036
hourglass international
HG1553
4.13 in. 10.5cm Large Case of 1000 Each for $179.47
19423034
hourglass international
HG1551
3.34 in. 8.5cm Small Case of 1000 Each for $179.47
19423037
hourglass international
HG1554
4.52 in. 11.5cm X-Large Case of 1000 Each for $179.47
19423035
hourglass international
HG1552
3.74 in. 9.5cm Medium Case of 1000 Each for $179.47
Description & Specifications
Specifications
Certifications ASTM, FDA
Length (English) 12 in.
Length (Metric) 30.5cm
Material Vinyl
Glove Cuff Style Beaded/Rolled
Latex Free Yes
Powdered Powder-free
Thickness (English) 4.33mil
  • Reduces the risk of Type I latex allergy
  • Powder-free; Smooth
  • With a beaded cuff
  • Clean Class 100 Compatible
  • Static dissipative ESD qualities
  • Double bagged
  • Available in sizes small to X-Large
Meet or exceed ASTM and FDA requirements