missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CBS Scientific Horizontal Standard Mini-Gel Kits

Used with Mini-Gel Horizontal Systems

Manufacturer:  CBS Scientific MHU202

 View more versions of this product

Catalog No. S22878



Description

Description

System features platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.
  • Platinum electrode cassettes can be easily removed for cleaning or replacement
  • High quality, heavy duty construction with vast array of accessory options
  • Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
  • Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
  • Tapered baffles allow easy in situ gel formation with or without gel tray
  • Run up to 32 samples per gel. Two slots in each gel tray
  • Combs are printed with catalog number and thickness for easy identification
  • Choice of double-sided adjustable height combs gives researcher more options
  • Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
  • Available in four vivid colors; green, yellow, blue or fluorescent pink
  • Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Applications: Seperation and identification of DNA fragments, RNA fragments, PCR Products and synthetic oligonucleotides
Specifications

Specifications

CE
Compatible Gel Tray: MGT-122
20.5 x 10.5 x 13 cm (8.0 x 4.1 x 5.1 in.)
L x W: 10 x 7.7 cm (3.9 x 3.0 in.)
3 Years
Standard Clear
Gel Tray I.D.: 10 x 7.2 cm (3.9 x 2.8 in.)
Mini-Gel Kit
250 mL
SDS
Documents
Product Certifications