missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CBS Scientific Horizontal Mini-Gel Productivity Packages

Allows casting second gel while first is running for increased efficiency.

Manufacturer:  CBS Scientific GCMHU202

 View more versions of this product

Catalog No. S2872



Description

Description

CBS Mini Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.
  • Platinum electrode cassettes can be easily removed for cleaning or replacement
  • High quality, heavy duty construction with vast array of accessory options
  • Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
  • Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
  • Tapered baffles allow easy in situ gel formation with or without gel tray
  • Run up to 32 samples per gel. Two slots in each gel tray
  • Combs are printed with catalog number and thickness for easy identification
  • Choice of double-sided adjustable height combs gives researcher more options
  • Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
  • Horizontal Mini-Gel Systems are available in four vivid colors; green, yellow, blue or fluorescent pink
  • Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Applications: Seperation and identification of DNA fragments, RNA fragments, PCR Products and synthetic oligonucleotides
Specifications

Specifications

CE
Compatible Gel Tray: MGT-122
20.5 x 10.5 x 13 cm (8.0 x 4.1 x 5.1 in.)
L x W: 10 x 7.7 cm (3.9 x 3.0 in.)
3 Years
Standard Clear
Gel Tray I.D.: 10 x 7.2 cm (3.9 x 2.8 in.)
Mini-Gel System
250 mL
SDS
Documents
Product Certifications