Horizontal Mini-Gel Productivity Kit
Increase productivity with external gel caster with extra comb and gel tray.
$454.95 - $454.95
Specifications
For Use With (Application) Separating and identifying DNA and RNA fragments, PCR products, and synthetic oligonucleotides
View More Specs
Includes: Horizontal unit, external gel caster with extra comb and gel tray, tapered baffles for submarine gel formation, safety cover with attached power leads and two UVT gel trays (each with two sides for two comb positions). Dual-sided combs available for 5.5W × 8.5cmL (2.1 × 3.3 in. ) and 7.5W × 10cmL (2.9 × 3.9 in. ); other units have single-sided adjustable height combs.
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S05749 Fisher Science Education
GCMGU-202
Each for $454.95
Specifications
Description
For use in separating and identifying DNA and RNA fragments, PCR products and synthetic oligonucleotides.
Certifications
CE marked.
Description & Specifications
Specifications
Separating and identifying DNA and RNA fragments, PCR products, and synthetic oligonucleotides