Troemner™ Hooked Stainless-Steel Weight Sets and Individual Weights
Weights can be hooked together at top and bottom.
Manufacturer:  Troemner™ 50500S
 View more versions of this product
Catalog No. S40307A
Description
Description
The design of the hook weight allows the bottom hook to be recessed in the bottom so the hook weights can be set on a flat surface.
  • Adjusted to tolerances specified by ANSI™/ASTM™ Class 7
  • Weight sets are supplied in an open plastic storage case
Specifications
Specifications
Metric hook weight
ANSI/ASTM Class 7
500g
Education
Stainless Steel
7.85g/cm3 at 20°C
Documents