Learn More
Honokiol, 98+%, Thermo Scientific Chemicals
Induces apoptosis in human squamous lung cancer cells
Supplier: Thermo Scientific Chemicals J63434MC
Description
Induces apoptosis in human squamous lung cancer cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Honokiol | |
86°C | |
C18H18O2 | |
100 mg | |
14,4742 | |
FVYXIJYOAGAUQK-UHFFFAOYSA-N | |
2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol | |
72303 | |
266.34 | |
Powder |
35354-74-6 | |
White | |
MFCD00016674 | |
UN3077 | |
honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl | |
C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C | |
266.34 | |
CHEBI:5759 | |
≥98% |
Chemical Identifiers
35354-74-6 | |
266.34 | |
FVYXIJYOAGAUQK-UHFFFAOYSA-N | |
72303 | |
2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol |
C18H18O2 | |
MFCD00016674 | |
honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl | |
CHEBI:5759 | |
C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only