Honeywell™ Whiting & Davis™ Metal Mesh Gloves
Constructed of individually welded and joined stainless steel rings
Manufacturer:  Honeywell™ A515XLD
 View more versions of this product
Catalog No. 19-069892
Description
Description
  • Individually welded rings allow the mesh fabric to flow without harsh edges
  • Seams are 4-ring joined, sealing the seam so gaps don't exist
  • Thumbs are attached with a 3-ring joint for a strong seam at a high wear area
  • Ergonomically designed and constructed with tapered fingers and palms to prevent sagging
  • Gloves are 3-step reversible to be worn on either hand
  • Mesh is manufactured following the European Standard of EN1082
Order Info
Sold by the glove.
Specifications
Specifications
Ambidextrous
Strap with dome fastener
Stainless Steel
Silver
Stainless Steel Mesh
food service, knife and blade sharpening/cleaning, fishing, garment manufacturing, veterinary services, animal control, Paper and pulp industry
X-Large