Honeywell™ Whiting & Davis™ Metal Mesh Gloves
Constructed of individually welded and joined stainless steel rings
Manufacturer:  Honeywell™ A515SD
 View more versions of this product
Catalog No. 19-069884
Description
Description
  • Individually welded rings allow the mesh fabric to flow without harsh edges
  • Seams are 4-ring joined, sealing the seam so gaps don't exist
  • Thumbs are attached with a 3-ring joint for a strong seam at a high wear area
  • Ergonomically designed and constructed with tapered fingers and palms to prevent sagging
  • Gloves are 3-step reversible to be worn on either hand
  • Mesh is manufactured following the European Standard of EN1082
Order Info
Sold by the glove.
Specifications
Specifications
Stainless Steel Mesh
food service, knife and blade sharpening/cleaning, fishing, garment manufacturing, veterinary services, animal control, Paper and pulp industry
Small
18 in.
Strap with dome fastener
Stainless Steel
Silver
45.72 cm