Learn More
Honeywell Safety Products™ Saline Wall Station
The Honeywell Saline Wall Station is easily mounted in hazardous areas to provide immediate relief in case of eye injury.
$67.28 - $476.39
Specifications
Certifications/Compliance | FDA |
---|---|
For Use With (Application) | Flushing Nuisance Particles Such as Pollen, Dust, Saw Dust, Smoke and More |
For Use With (Equipment) | For Refill Bottles, See 19-003-030. |
Material | Polyethylene |
Alert:
Since the first few seconds of an eye injury are critical, an eyewash bottle station will provide immediate relief until the worker has access to either a plumbed station or a shower. Eyewash bottles support a plumbed or self-contained drenching facility. They do not take the place of a 15-minute flushing facility.
Catalog Number | Mfr. No. | Capacity (English) | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Product Type | Price | Quantity | ||||
19-085-598
|
Honeywell Safety Products™
320004620000H5 |
32 oz. | Double Eye Wall Station |
Case of 4 Each for $320.63
|
|
||||
19-003-029
|
Honeywell Safety Products™
320004600000H5 |
16 oz. | Single Eye Wall Station |
Case of 8 Each for $476.39
|
|
||||
19-003-031
|
Honeywell Safety Products™
320004610000 |
32 oz. | Single Eye Wall Station |
Each for $67.28
Case of 8 Each for $497.44
|
|
||||
Description
The Honewell Saline Wall Station is easily mounted in hazardous areas to provide immediate relief in case of eye injury.
Choose from 16 or 32 oz. (473 or 946mL) bottles.
- Highly visible, with full emergency-use instructions on front
- Made from sturdy, high-impact plastic
- Bottles are single-use only with snap-off tops for quick and easy access
- Bottles have extended flow nozzles; 32 oz. bottles have integral grip for ease of use in an emergency
Order Info
Refill bottles sold separately. See Cat. Nos. 19-003-030 and -033.
Compliance
Does not meet the criteria of plumbed or self-contained eyewash equipment.
Specifications
FDA | |
Flushing Nuisance Particles Such as Pollen, Dust, Saw Dust, Smoke and More | |
For Refill Bottles, See 19-003-030. | |
Polyethylene |