Please login to your online account to display your discounted pricing
Honeywell Fendall Saline Cartridge for Pureflow 1000 and 2000 Eyewash Stations
Sterile Saline Cartridge for Sperian Fendall Pureflow 1000 and 2000 Emergency Eyewash Stations
$360.24 - $360.24
Specifications
Includes Nozzle assembly and delivery hose
Shelf Life 2 Years
View More Specs
Includes:  Nozzle assembly and delivery hose
Products
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity    
19-175-434
honeywell
32ST10500000
Fendall 1000 portable emergency eyewash station Saline Cartridge Each of 2 for $360.24
Description & Specifications
Specifications
Includes Nozzle assembly and delivery hose
Shelf Life 2 Years
  • Each cartridge contains 6.87 gal. (26L) of 100% sterile saline solution
  • 24-month cartridge life reduces the cost of replacement and saves inspection time; cartridge expiration date is easily viewed through side windows of eyewash unit
Safety and Handling
2 Years