Please login to your online account to display your discounted pricing

Honeywell Premium Natural Rubber Latex Gloves

$451.31 - $451.31

Specifications

Certifications 21 CFR, ANSI, ISEA 105, ASTM D 3389-05, F1790-97, USDA, FDA
Color Orange
Color Yellow
Length (English) 15 in.
Length (Metric) 38.1cm
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-150-1074

 
honeywell
PNLB1815/O/8
8 Case of 144 Each for $451.31

19-150-1076

 
honeywell
PNLB1815/O/10
10 Case of 144 Each for $451.31
Description & Specifications

Specifications

Certifications 21 CFR, ANSI, ISEA 105, ASTM D 3389-05, F1790-97, USDA, FDA
Color Orange
Color Yellow
Length (English) 15 in.
Length (Metric) 38.1cm
Material Rubber, natural
Exterior Finish Embossed, Diamond
For Use With (Application) automotive assembly, chemical handling and processing, food processing and preparation, janitorial
Glove Cuff Style Beaded/Rolled
Powdered Powder-free
Thickness (English) 18mil