missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ Pure Fit PF570 HPPE Cut-Resistant Gloves, Nitrile Coated
Nitrile coated ANSI Cut Level 4 protection gloves offer dexterity and tactile sensitivity for work requiring precision handling. Seamless knit construction eliminates uncomfortable seams that can rub or chafe.
$43.34 - $43.34
Specifications
ANSI Abrasion Rating | ANSI 4 |
---|---|
Coating Material | Nitrile |
Color | Gray,Light Gray |
Cut Resistance Rating | ANSI A5 |
Material | HPPE,Stainless Steel |
Catalog Number | Mfr. No. | Size | Glove Cuff Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Glove Cuff Color | Price | Quantity | ||||
19-130-5800
|
Honeywell™
PF570M |
Medium | Blue |
Pair for $43.34
Dozen of 12 Pair for $504.60 |
|
Description
- White high performance polyethylene (HPPE) and stainless steel blended shell
- Lycra™ plating provides comfortable, snug fit
- Gray nitrile palm and fingertip coating provides extra durability and secure grip
- 13-gauge, lightweight construction
Specifications
ANSI 4 | |
Gray,Light Gray | |
HPPE,Stainless Steel | |
EN 388 | |
Pure Fit PF570 |
Nitrile | |
ANSI A5 | |
Cut Resistant Gloves | |
Precision handling |