Please login to your online account to display your discounted pricing

Honeywell Perfect Fit Searchlight Liner Gloves

$40.42 - $40.42

Specifications

Color Yellow
Cut Resistance Rating Level 2
Material Kevlar™
For Use With (Application) light duty applications
Weight Light weight
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-816-549

 
honeywell
KV13A
Men's Pack of 12 Pair for $40.42
Description & Specifications

Specifications

Color Yellow
Cut Resistance Rating Level 2
Material Kevlar™
For Use With (Application) light duty applications
Weight Light weight