Please login to your online account to display your discounted pricing

Honeywell Perfect Fit™ Cut-Resistant Gloves

Dyneema fiber gloves provide a high level of cut protection in a lightweight glove

$140.41 - $195.03

Specifications

Cut Resistance Rating Level 5
Material HPPE (High-performance Polyethylene)
For Use With (Application) food processing, food packaging, meat and poultry industries, restaurants, commercial fishing and canning
Hand Ambidextrous
View More Specs
Products
Catalog Number Mfr. No. Style Color Size Price Quantity    

19049895D

 
honeywell
PF13-L
13-Cut White Large Dozen of 12 Each for $140.41

29-635-7881

 
honeywell
PF13GYMSSPL
13-Cut Gray Medium Case of 12 Each for $187.18

19-130-5798

 
north safety products
PF13GYLSSPL
13-Cut White Large Dozen of 12 Each for $195.03
Description & Specifications

Specifications

Cut Resistance Rating Level 5
Material HPPE (High-performance Polyethylene)
For Use With (Application) food processing, food packaging, meat and poultry industries, restaurants, commercial fishing and canning
Hand Ambidextrous

Lightweight, low-linting and does not easily absord liquids; these gloves are excellent for food processing applications

  • Color-coded PowerCuff™ helps to eliminate cuff “blow-out” from repeated laundering
  • Can easily be sorted by size
  • Sold individually
  • Complies with USDA and FDA regulations, 21 CFR, for use in food processing