Honeywell™ Perfect Fit™ Cut-Resistant Gloves
Dyneema fiber gloves provide a high level of cut protection in a lightweight glove
Manufacturer:  Honeywell™ PF13GYMSSPL
 View more versions of this product
Catalog No. 29-635-7881
Description
Description
Lightweight, low-linting and does not easily absord liquids; these gloves are excellent for food processing applications
  • Color-coded PowerCuff™ helps to eliminate cuff “blow-out” from repeated laundering
  • Can easily be sorted by size
  • Sold individually
  • Complies with USDA and FDA regulations, 21 CFR, for use in food processing
Specifications
Specifications
Safety
Can be machine laundered using commercial detergents (up to pH 15), water temperatures up to 49°C (120°F,) and drying temperatures up to 82°C (180°F.) HPPE can be sanitized and bleached without affecting performance. WARNING : Laundering gloves that have been in contact with toxic or corrosive chemicals may result in a chemical reaction, including fumes that could cause person al injury, serious illness or death.
Yellow
HPPE
13-Cut
Gray
Level 5
Ambidextrous
food processing, food packaging, meat and poultry industries, restaurants, commercial fishing and canning
Medium
Light weight
Yellow
Documents