Please login to your online account to display your discounted pricing

Honeywell NorthFlex™ Nitri Task KL Gloves

Comfortable, dexterous cut-resistant work gloves

$90.03 - $90.03

Specifications

Coated Area Palm, finger
Color Black
Color Yellow
Cut Resistance Rating Level 2
Material Nitrile
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-150-0993

 
honeywell
NFKL13/7S
Small Dozen of 12 Each for $90.03
Description & Specifications

Specifications

Coated Area Palm, finger
Color Black
Color Yellow
Cut Resistance Rating Level 2
Material Nitrile
Exterior Finish Textured
For Use With (Application) cut and abrasion resistance without compromising dexterity, tactility and comfort
Glove Cuff Style Knit

  • Yellow, 13 gauge seamless Kevlar™/Lycra™ liner for cut resistance and comfort
  • Black nitrile coating covers palm and fingertips and is textured for added grip
  • Can be exposed to solvents, oils and fats
  • Knitted cuff with edge color-coded by size
  • Latex-free

Construction, manufacturing, municipal services, oil and gas

Limited