Honeywell™ NitriTask Supported Foam Nitrile Gloves
Versatile cut-resistant work gloves
Manufacturer:  Honeywell™ NFF13SZ8
 View more versions of this product
Catalog No. 19-130-1657
Description
Description
  • Nylon seamless liner (grey)
  • Foam nitrile palm coating (grey)
  • Knitted wrist
  • Automotive assembly
  • Maintenance work
  • Light assembly
  • Manufacturing
  • Small part handling
Specifications
Specifications
Safety
Launderable
automotive assembly, general maintenance work
White Nylon
Gray
2.54 cm
Level 1
Knit
Nitrile
8
1 in.
Documents