Please login to your online account to display your discounted pricing

Honeywell NitriTask Supported Foam Nitrile Gloves

Versatile cut-resistant work gloves

$46.73 - $46.73

Specifications

Cleaning Procedure Launderable
Color Gray
Cut Resistance Rating Level 1
Length (English) 1 in.
Length (Metric) 2.54cm
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-130-1659

 
honeywell
NFF1310XL
10 Dozen of 12 Pair for $46.73
Case of 12 Dozen for $512.04

19-130-1658

 
honeywell
NFF13SZ9
9 Dozen of 12 Pair for $46.73

19-130-1657

 
honeywell
NFF13SZ8
8 Dozen of 12 Pair for $46.73
Description & Specifications

Specifications

Cleaning Procedure Launderable
Color Gray
Cut Resistance Rating Level 1
Length (English) 1 in.
Length (Metric) 2.54cm
Material Nitrile
For Use With (Application) automotive assembly, general maintenance work
Glove Cuff Style Knit

  • Nylon seamless liner (grey)
  • Foam nitrile palm coating (grey)
  • Knitted wrist

  • Automotive assembly
  • Maintenance work
  • Light assembly
  • Manufacturing
  • Small part handling