Please login to your online account to display your discounted pricing

Honeywell Long Sleeve PVC Gloves

$54.43 - $54.43

Specifications

Certifications ANSI/ISEA 105, ASTM D 3389-05, ANSI/ISEA 105, ASTM F 1790-97, EN 374, EN 388, EN 420, OSHA 29 CFR 1915.157
Color Green
Length (English) 26 in.
Length (Metric) 66.04cm
Material PVC (Polyvinyl Chloride)
View More Specs
Products
Catalog Number Mfr. No. Glove Cuff Style Price Quantity    

19-150-1012

 
honeywell
1812FWG24
Elastic Each for $54.43
Description & Specifications

Specifications

Certifications ANSI/ISEA 105, ASTM D 3389-05, ANSI/ISEA 105, ASTM F 1790-97, EN 374, EN 388, EN 420, OSHA 29 CFR 1915.157
Color Green
Length (English) 26 in.
Length (Metric) 66.04cm
Material PVC (Polyvinyl Chloride)
Size Large
Exterior Finish Rough
For Use With (Application) agriculture, chemical and food services
Hand Ambidextrous
Lining Interlock knit