Please login to your online account to display your discounted pricing
Honeywell Grip-Task Series Natural Rubber Gloves
Protect against abrasion, liquids and chemicals
$5.57 - $355.50
Products
Catalog Number Mfr. No. Price Quantity    
19-150-0960
honeywell
T65FWG/11XXL
Case of 48 Each for $355.50
29-635-5765
honeywell
T66FWG11XXL
Pair for $5.57
Dozen of 12 Pair for $63.96
Description & Specifications
  • Natural rubber gloves with rough finish natural rubber coating for added grip on wet surfaces
  • Available with gauntlet or safety style cuffs
  • Liquid proof
  • Chemical resistant
  • 30cm (12 in.) long
Chemicals, construction, manufacturing, municipal services
EN374 - Permiation and breakthrough time; EN388 - Mechanical risks from abrasion, cut, tear and puncture; EN420 - General Requirements; OSHA Standard -29 CFR 1915.157
Limited