missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Flex Nitri Task C5 Foamed Nitrile Palm Coated Aramid/Steel Gloves

$19.80 - $19.80

Specifications

Color Dark Blue
Material Aramid,Steel
Sterility Sterile
For Use With (Application) sheet Metal fabrication, machine shops, Glass industry, automotive and other assembly work
Glove Cuff Style Knit
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-130-270A
View Documents
Honeywell™
NFF13C7S
Small
Pair for $19.80
Pack of 12 Pair for $227.16
 
19-130-270B
View Documents
Honeywell™
NFF13C8M
Medium
Pair for $19.80
Pack of 12 Pair for $227.16
 
19-130-270C
View Documents
Honeywell™
NFF13C9L
Large
Pair for $19.80
Pack of 12 Pair for $227.16
 
19-130-270D
View Documents
Honeywell™
NFF13C10XL
X-Large
Pair for $19.80
Pack of 12 Pair for $227.16
 
Description

Description

Specifications

Specifications

Dark Blue
Sterile
Knit
Aramid,Steel
sheet Metal fabrication, machine shops, Glass industry, automotive and other assembly work
SDS