Please login to your online account to display your discounted pricing

Honeywell Eco Knit Gloves

Cotton/polyester knit glove for general purpose hand protection

$132.58 - $132.58

Specifications

Certifications CE, EN 420, USDA, FDA, 21 CFR
Color Natural
Material Cotton
Material Polyester
For Use With (Application) food handling, janitorial, light assembly, small part handling, shipping/receiving
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-150-0900

 
honeywell
11RK/7S
Small Case of 240 Each for $132.58

19-150-0901

 
honeywell
11RK/8M
Medium Case of 240 Each for $132.58

19-150-0902

 
honeywell
11RK/9L
Large Case of 240 Each for $132.58
Description & Specifications

Specifications

Certifications CE, EN 420, USDA, FDA, 21 CFR
Color Natural
Material Cotton
Material Polyester
For Use With (Application) food handling, janitorial, light assembly, small part handling, shipping/receiving
Glove Cuff Style Eco knit
Hand Ambidextrous

  • Seamless knit design
  • Ambidextrous
  • No seams to rub or chafe for added wearer comfort
  • 80% polyester, 20% cotton

Complies with USDA and FDA regulations, 21 CFR, for use in food processing