Honeywell Dexi-Task Nitrile Gloves
For industrial examination and medical use
$161.38 - $191.77
Specifications
Certifications USDA, FDA, 21 CFR, EN 455-1, 2, EN 374-1, EN 420, ASTM D 3578
Color Blue
Length (English) 9 in.
Length (Metric) 23cm
Material Nitrile
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Grade Width (English) Powdered Size Price Quantity  
Catalog Number Mfr. No. Grade Width (English) Powdered Size Price Quantity  
19-150-1038 Honeywell
LA049IND/S
Industrial 3.3 in. Powdered Small Case of 1000 Each for $161.38
19-150-1035 Honeywell
LA049PF/M
Medical 3.74 in. Powder-free Medium Case of 1000 Each for $191.77
19-150-1037 Honeywell
LA049PF/XL
Medical 4.5 in. Powder-free X-Large Case of 1000 Each for $191.77
19-150-1036 Honeywell
LA049PFL
Medical 4.13 in. Powder-free Large Case of 1000 Each for $191.77
19-150-1034 Honeywell
LA049PF/S
Medical 3.3 in. Powder-free Small Case of 1000 Each for $191.77
19-150-1039 Honeywell
LA049INDM
Industrial 3.74 in. Powdered Medium Case of 1000 Each for $161.38
19-150-1040 Honeywell
LA049IND/L
Industrial 4.13 in. Powdered Large Case of 1000 Each for $161.38
19-150-1041 Honeywell
LA049IND/XL
Industrial 4.5 in. Powdered X-Large Case of 1000 Each for $161.83
Specifications
Description
  • Puncture resistant
  • 5mil thickness
  • Blue
Certifications
510K approved for medical use
Description & Specifications
Specifications
USDA, FDA, 21 CFR, EN 455-1, 2, EN 374-1, EN 420, ASTM D 3578
9 in.
Nitrile
Rolled
Yes
5 mil
Blue
23cm
Yes
Ambidextrous
16