Honeywell™ North™ Silver Shield™ Chemical Resistant Apparel
Various chemical resistant apparel
Manufacturer: Honeywell™ HNYSSS
Description
- Five layer PE and EVOH constructions provides excellent chemical resistant to over 280 different chemicals including alchohols, aliphatics, aromatice, chlorines, ketones and esters
- Provides high performance protection in the most demanding applications
Specifications
Elastic | |
Silver | |
Universal |
Sleeves | |
Composite Fabric |